4-(2,5-Dimethylphenoxy)piperidine hydrochloride

Supplier: Matrix Scientific

023278-500MG
101815-692EA 176.92 USD
101815-692
4-(2,5-Dimethylphenoxy)piperidine hydrochloride
4-(2,5-Dimethylphenoxy)piperidine hydrochloride

Hazard: Irritant

Formula: C₁₃H₂₀ClNO
MDL Number: MFCD11100497
CAS Number: 1185002-20-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR