4-(2,4-Dimethylphenoxy)piperidine hydrochloride

Supplier: Matrix Scientific

023277-500MG
101815-690EA 402.5 USD
101815-690
4-(2,4-Dimethylphenoxy)piperidine hydrochloride
4-(2,4-Dimethylphenoxy)piperidine hydrochloride

Hazard: Irritant

Formula: C₁₃H₂₀ClNO
MDL Number: MFCD11845774
CAS Number: 1185039-63-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR