4-(2,3,5-Trimethylphenoxy)piperidine hydrochloride

Supplier: Matrix Scientific

023276-500MG
101815-688EA 538.46 USD
101815-688
4-(2,3,5-Trimethylphenoxy)piperidine hydrochloride
4-(2,3,5-Trimethylphenoxy)piperidine hydrochloride

Hazard: Irritant

Formula: C₁₄H₂₂ClNO
MDL Number: MFCD11845773
CAS Number: 1184976-36-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR