4-([1,1'-Biphenyl]-2-yloxy)piperidine
Supplier: Matrix Scientific
023273-500MG
101815-680EA
315.38
USD
101815-680
4-([1,1'-Biphenyl]-2-yloxy)piperidine
4-([1,1'-Biphenyl]-2-yloxy)piperidine
Hazard: Irritant
Formula:
C₁₇H₁₉NO |
MDL Number:
MFCD06245999 CAS Number: 883515-53-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...