4-(2-Ethoxyphenoxy)piperidine

Supplier: Matrix Scientific

023272-500MG
101815-678EA 315.38 USD
101815-678
4-(2-Ethoxyphenoxy)piperidine
4-(2-Ethoxyphenoxy)piperidine

Hazard: Irritant

Formula: C₁₃H₁₉NO₂
MDL Number: MFCD08061063
CAS Number: 420137-19-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR