4-(2-Methoxyphenoxy)piperidine hydrochloride

Supplier: Matrix Scientific

Synonyms: 1-Methoxy-2-[(piperidin-4-yl)oxy]benzene hydrochloride2-[(Piperidin-4-yl)oxy]anisole hydrochloride

023271-500MG
101815-676EA 315.38 USD
101815-676
4-(2-Methoxyphenoxy)piperidine hydrochloride
4-(2-Methoxyphenoxy)piperidine hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₇NO₂·ClH
MW: 243.73 g/mol
MDL Number: MFCD03840131
CAS Number: 6024-31-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR