4-[(2-Methyl-2-propenyl)oxy]piperidine hydrochloride

Supplier: Matrix Scientific

023264-500MG
101815-662EA 495.38 USD
101815-662
4-[(2-Methyl-2-propenyl)oxy]piperidine hydrochloride
4-[(2-Methyl-2-propenyl)oxy]piperidine hydrochloride

Hazard: Irritant

Formula: C₉H₁₈ClNO
MDL Number: MFCD11100496
CAS Number: 1185077-23-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR