4-([1,1'-Biphenyl]-4-ylmethoxy)piperidine hydrochloride

Supplier: Matrix Scientific

023261-500MG
101815-656EA 284.62 USD
101815-656
4-([1,1'-Biphenyl]-4-ylmethoxy)piperidine hydrochloride
4-([1,1'-Biphenyl]-4-ylmethoxy)piperidine hydrochloride

Hazard: Irritant

Formula: C₁₈H₂₂ClNO
MDL Number: MFCD09879218
CAS Number: 1185298-30-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR