4-[(4-Fluorobenzyl)oxy]piperidine hydrochloride

Supplier: Matrix Scientific

023260-500MG
101815-654EA 315.38 USD
101815-654
4-[(4-Fluorobenzyl)oxy]piperidine hydrochloride
4-[(4-Fluorobenzyl)oxy]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₇ClFNO
MW: 245.73 g/mol
MDL Number: MFCD11506395
CAS Number: 1185107-71-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR