4-[(3-Methylbenzyl)oxy]piperidine hydrochloride

Supplier: Matrix Scientific

023259-500MG
101815-652EA 284.62 USD
101815-652
4-[(3-Methylbenzyl)oxy]piperidine hydrochloride
4-[(3-Methylbenzyl)oxy]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₃H₂₀ClNO
MDL Number: MFCD09879217
CAS Number: 1185304-38-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR