4-[(3-Chlorobenzyl)oxy]piperidine hydrochloride

Supplier: Matrix Scientific

023258-500MG
101815-650EA 315.38 USD
101815-650
4-[(3-Chlorobenzyl)oxy]piperidine hydrochloride
4-[(3-Chlorobenzyl)oxy]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₇Cl₂NO
MDL Number: MFCD09879216
CAS Number: 1185020-93-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR