4-[(3-Fluorobenzyl)oxy]piperidine hydrochloride

Supplier: Matrix Scientific

023257-500MG
101815-648EA 325.45 USD
101815-648
4-[(3-Fluorobenzyl)oxy]piperidine hydrochloride
4-[(3-Fluorobenzyl)oxy]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₇ClFNO
MW: 245.73 g/mol
MDL Number: MFCD11506394
CAS Number: 1185298-16-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR