4-[(2-Methylbenzyl)oxy]piperidine hydrochloride

Supplier: Matrix Scientific

023256-500MG
101815-646EA 435.38 USD
101815-646
4-[(2-Methylbenzyl)oxy]piperidine hydrochloride
4-[(2-Methylbenzyl)oxy]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₃H₂₀ClNO
MDL Number: MFCD09879215
CAS Number: 1185175-54-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR