4-[(2-Methoxybenzyl)oxy]piperidine hydrochloride

Supplier: Matrix Scientific

023255-500MG
101815-644EA 338.46 USD
101815-644
4-[(2-Methoxybenzyl)oxy]piperidine hydrochloride
4-[(2-Methoxybenzyl)oxy]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₃H₂₀ClNO₂
MDL Number: MFCD09997937
CAS Number: 81151-57-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR