4-Ethoxypiperidine

Supplier: Matrix Scientific

023253-500MG 023253-2.5G
101815-642EA 2141.37 USD
101815-642 101815-640
4-Ethoxypiperidine
4-Ethoxypiperidine

Hazard: Irritant

Formula: C₇H₁₅NO
MDL Number: MFCD06247385
CAS Number: 1122-86-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR