4-(Isopentyloxy)piperidine hydrochloride

Supplier: Matrix Scientific

023251-500MG
101815-636EA 538.46 USD
101815-636
4-(Isopentyloxy)piperidine hydrochloride
4-(Isopentyloxy)piperidine hydrochloride

Hazard: Irritant

Formula: C₁₀H₂₂ClNO
MDL Number: MFCD09800637
CAS Number: 1172347-43-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR