4-(Allyloxy)piperidine hydrochloride
Supplier: Matrix Scientific
023250-500MG
101815-634EA
284.62
USD
101815-634
4-(Allyloxy)piperidine hydrochloride
4-(Allyloxy)piperidine hydrochloride
Hazard: Irritant
Formula:
C₈H₁₆ClNO |
MDL Number:
MFCD11506393 CAS Number: 956324-63-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...