4-[(3-Methyl-2-butenyl)oxy]piperidine hydrochloride

Supplier: Matrix Scientific

023249-500MG
101815-632EA 235.75 USD
101815-632
4-[(3-Methyl-2-butenyl)oxy]piperidine hydrochloride
4-[(3-Methyl-2-butenyl)oxy]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₀H₂₀ClNO
MDL Number: MFCD12026504
CAS Number: 1185044-73-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR