3-(4-Fluoro-2-methylphenoxy)piperidine

Supplier: Matrix Scientific

023244-500MG
101815-622EA 315.38 USD
101815-622
3-(4-Fluoro-2-methylphenoxy)piperidine
3-(4-Fluoro-2-methylphenoxy)piperidine

Hazard: Irritant

Formula: C₁₂H₁₆FNO
MDL Number: MFCD08687863
CAS Number: 946759-56-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR