3-(4-Chloro-2-fluorophenoxy)piperidine

Supplier: Matrix Scientific

023243-500MG
101815-620EA 315.38 USD
101815-620
3-(4-Chloro-2-fluorophenoxy)piperidine
3-(4-Chloro-2-fluorophenoxy)piperidine

Hazard: Irritant

Formula: C₁₁H₁₃ClFNO
MDL Number: MFCD08687326
CAS Number: 946726-21-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR