3-(2-Chloro-5-fluorophenoxy)piperidine

Supplier: Matrix Scientific

023240-500MG
101815-614EA 235.75 USD
101815-614
3-(2-Chloro-5-fluorophenoxy)piperidine
3-(2-Chloro-5-fluorophenoxy)piperidine

Hazard: Irritant

Formula: C₁₁H₁₃ClFNO
MDL Number: MFCD08687324
CAS Number: 946759-52-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR