4-(2-Methoxyethyl)phenyl-3-piperidinyl ether

Supplier: Matrix Scientific

023238-500MG
101815-610EA 315.38 USD
101815-610
4-(2-Methoxyethyl)phenyl-3-piperidinyl ether
4-(2-Methoxyethyl)phenyl-3-piperidinyl ether

Hazard: Irritant

Formula: C₁₄H₂₁NO₂
MDL Number: MFCD08688045
CAS Number: 946681-36-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR