3-(3-Ethoxyphenoxy)piperidine

Supplier: Matrix Scientific

023237-500MG
101815-608EA 235.75 USD
101815-608
3-(3-Ethoxyphenoxy)piperidine
3-(3-Ethoxyphenoxy)piperidine

Hazard: Irritant

Formula: C₁₃H₁₉NO₂
MDL Number: MFCD08687741
CAS Number: 946714-65-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR