2-Naphthyl-3-piperidinyl ether

Supplier: Matrix Scientific

023225-500MG
101815-584EA 315.38 USD
101815-584
2-Naphthyl-3-piperidinyl ether
2-Naphthyl-3-piperidinyl ether

Hazard: Irritant

Formula: C₁₅H₁₇NO
MDL Number: MFCD08688440
CAS Number: 946726-09-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR