3-[(4-Chloro-1-naphthyl)oxy]piperidine hydrochloride

Supplier: Matrix Scientific

023224-500MG
101815-582EA 538.46 USD
101815-582
3-[(4-Chloro-1-naphthyl)oxy]piperidine hydrochloride
3-[(4-Chloro-1-naphthyl)oxy]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₅H₁₇Cl₂NO
MDL Number: MFCD12405062
CAS Number: 946714-56-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR