2,3-Dihydro-1H-inden-5-yl-3-piperidinyl ether

Supplier: Matrix Scientific

023222-500MG
101815-578EA 235.75 USD
101815-578
2,3-Dihydro-1H-inden-5-yl-3-piperidinyl ether
2,3-Dihydro-1H-inden-5-yl-3-piperidinyl ether

Hazard: Irritant

Formula: C₁₄H₁₉NO
MDL Number: MFCD08687599
CAS Number: 946759-36-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR