3-(4-Ethylphenoxy)piperidine

Supplier: Matrix Scientific

023221-500MG
101815-576EA 315.38 USD
101815-576
3-(4-Ethylphenoxy)piperidine
3-(4-Ethylphenoxy)piperidine

Hazard: Irritant

Formula: C₁₃H₁₉NO
MDL Number: MFCD08687784
CAS Number: 946726-06-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR