3-(4-Benzylphenoxy)piperidine

Supplier: Matrix Scientific

023219-500MG
101815-574EA 315.38 USD
101815-574
3-(4-Benzylphenoxy)piperidine
3-(4-Benzylphenoxy)piperidine

Hazard: Irritant

Formula: C₁₈H₂₁NO
MDL Number: MFCD08686907
CAS Number: 946681-24-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR