3-[4-(sec-Butyl)phenoxy]piperidine

Supplier: Matrix Scientific

023216-500MG
101815-568EA 315.38 USD
101815-568
3-[4-(sec-Butyl)phenoxy]piperidine
3-[4-(sec-Butyl)phenoxy]piperidine

Hazard: Irritant

Formula: C₁₅H₂₃NO
MDL Number: MFCD08687203
CAS Number: 946759-32-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR