3-(4-Isopropylphenoxy)piperidine

Supplier: Matrix Scientific

023215-500MG
101815-566EA 235.75 USD
101815-566
3-(4-Isopropylphenoxy)piperidine
3-(4-Isopropylphenoxy)piperidine

Hazard: Irritant

Formula: C₁₄H₂₁NO
MDL Number: MFCD08688002
CAS Number: 946726-03-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR