3-[4-(Trifluoromethyl)phenoxy]piperidine

Supplier: Matrix Scientific

023212-500MG
101815-560EA 198.46 USD
101815-560
3-[4-(Trifluoromethyl)phenoxy]piperidine
3-[4-(Trifluoromethyl)phenoxy]piperidine

Hazard: Irritant

Formula: C₁₂H₁₄F₃NO
MDL Number: MFCD08061014
CAS Number: 902836-91-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR