3-([1,1'-Biphenyl]-4-yloxy)piperidine hydrochloride

Supplier: Matrix Scientific

023210-500MG
101815-556EA 235.75 USD
101815-556
3-([1,1'-Biphenyl]-4-yloxy)piperidine hydrochloride
3-([1,1'-Biphenyl]-4-yloxy)piperidine hydrochloride

Hazard: Irritant

Formula: C₁₇H₂₀ClNO
MDL Number: MFCD11100495
CAS Number: 1185071-92-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR