3-(4-Ethoxyphenoxy)piperidine
Supplier: Matrix Scientific
023209-500MG
101815-554EA
315.38
USD
101815-554
3-(4-Ethoxyphenoxy)piperidine
3-(4-Ethoxyphenoxy)piperidine
Hazard: Irritant
Formula:
C₁₃H₁₉NO₂ |
MDL Number:
MFCD08687742 CAS Number: 946714-47-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...