3-(4-Phenoxyphenoxy)piperidine

Supplier: Matrix Scientific

023206-500MG
101815-548EA 315.38 USD
101815-548
3-(4-Phenoxyphenoxy)piperidine
3-(4-Phenoxyphenoxy)piperidine

Hazard: Irritant

Formula: C₁₇H₁₉NO₂
MDL Number: MFCD08688469
CAS Number: 946681-18-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR