3-(4-Chloro-3,5-dimethylphenoxy)piperidine hydrochloride

Supplier: Matrix Scientific

023205-500MG
101815-546EA 315.38 USD
101815-546
3-(4-Chloro-3,5-dimethylphenoxy)piperidine hydrochloride
3-(4-Chloro-3,5-dimethylphenoxy)piperidine hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₉Cl₂NO
MDL Number: MFCD12405043
CAS Number: 1185302-79-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR