3-(3-Ethylphenoxy)piperidine hydrochloride

023201-500MG
101815-538EA 308.2 USD
101815-538
3-(3-Ethylphenoxy)piperidine hydrochloride
3-(3-Ethylphenoxy)piperidine hydrochloride

Hazard: Irritant

Formula: C₁₃H₂₀ClNO
MDL Number: MFCD11506391
CAS Number: 946759-21-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR