3-(3,5-Dimethylphenoxy)piperidine

Supplier: Matrix Scientific

023200-500MG
101815-536EA 315.38 USD
101815-536
3-(3,5-Dimethylphenoxy)piperidine
3-(3,5-Dimethylphenoxy)piperidine

Hazard: Irritant

Formula: C₁₃H₁₉NO
MDL Number: MFCD08687659
CAS Number: 946681-15-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR