3-(3,4-Dimethylphenoxy)piperidine

Supplier: Matrix Scientific

023199-500MG
101815-534EA 315.38 USD
101815-534
3-(3,4-Dimethylphenoxy)piperidine
3-(3,4-Dimethylphenoxy)piperidine

Hazard: Irritant

Formula: C₁₃H₁₉NO
MDL Number: MFCD08687658
CAS Number: 946725-92-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR