Methyl-3-(3-piperidinyloxy)benzoate hydrochloride

Supplier: Matrix Scientific

023195-500MG
101815-526EA 186.15 USD
101815-526
Methyl-3-(3-piperidinyloxy)benzoate hydrochloride
Methyl-3-(3-piperidinyloxy)benzoate hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₈ClNO₃
MDL Number: MFCD09997064
CAS Number: 1185297-89-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR