3-(3-Methoxyphenoxy)piperidine

Supplier: Matrix Scientific

023194-500MG
101815-524EA 235.75 USD
101815-524
3-(3-Methoxyphenoxy)piperidine
3-(3-Methoxyphenoxy)piperidine

Hazard: Irritant

Formula: C₁₂H₁₇NO₂
MDL Number: MFCD08060989
CAS Number: 902837-25-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR