3-(3,4-Dichlorophenoxy)piperidine

Supplier: Matrix Scientific

023192-500MG
101815-520EA 538.46 USD
101815-520
3-(3,4-Dichlorophenoxy)piperidine
3-(3,4-Dichlorophenoxy)piperidine

Hazard: Irritant

Formula: C₁₁H₁₃Cl₂NO
MDL Number: MFCD08687541
CAS Number: 946714-37-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR