3-(3-Chloro-4-fluorophenoxy)piperidine

Supplier: Matrix Scientific

023191-500MG
101815-518EA 538.46 USD
101815-518
3-(3-Chloro-4-fluorophenoxy)piperidine
3-(3-Chloro-4-fluorophenoxy)piperidine

Hazard: Irritant

Formula: C₁₁H₁₃ClFNO
MDL Number: MFCD08687325
CAS Number: 946759-15-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR