2-Allylphenyl-3-piperidinyl ether
Supplier: Matrix Scientific
023187-500MG
101815-510EA
315.38
USD
101815-510
2-Allylphenyl-3-piperidinyl ether
2-Allylphenyl-3-piperidinyl ether
Hazard: Irritant
Formula:
C₁₄H₁₉NO |
MDL Number:
MFCD08686774 CAS Number: 946714-33-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...