3-(2,4-Dimethylphenoxy)piperidine hydrochloride

Supplier: Matrix Scientific

023182-500MG
101815-500EA 315.38 USD
101815-500
3-(2,4-Dimethylphenoxy)piperidine hydrochloride
3-(2,4-Dimethylphenoxy)piperidine hydrochloride

Hazard: Irritant

Formula: C₁₃H₂₀ClNO
MDL Number: MFCD12405042
CAS Number: 1185297-39-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR