3-(2,5-Dichlorophenoxy)piperidine

Supplier: Matrix Scientific

023174-500MG
101815-484EA 235.75 USD
101815-484
3-(2,5-Dichlorophenoxy)piperidine
3-(2,5-Dichlorophenoxy)piperidine

Hazard: Irritant

Formula: C₁₁H₁₃Cl₂NO
MDL Number: MFCD08687540
CAS Number: 946759-06-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR