3-(2,4-Dichlorophenoxy)piperidine hydrochloride

Supplier: Matrix Scientific

023173-500MG
101815-482EA 315.38 USD
101815-482
3-(2,4-Dichlorophenoxy)piperidine hydrochloride
3-(2,4-Dichlorophenoxy)piperidine hydrochloride

Hazard: Irritant

Formula: C₁₁H₁₄Cl₃NO
MDL Number: MFCD09879208
CAS Number: 1185302-73-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR