3-(2,3-Dichlorophenoxy)piperidine

Supplier: Matrix Scientific

023171-500MG
101815-478EA 235.75 USD
101815-478
3-(2,3-Dichlorophenoxy)piperidine
3-(2,3-Dichlorophenoxy)piperidine

Hazard: Irritant

Formula: C₁₁H₁₃Cl₂NO
MDL Number: MFCD08687538
CAS Number: 946714-21-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR