3-Phenoxypiperidine hydrochloride

Supplier: Matrix Scientific

023168-500MG
101815-472EA 315.38 USD
101815-472
3-Phenoxypiperidine hydrochloride
3-Phenoxypiperidine hydrochloride

Hazard: Irritant

Formula: C₁₁H₁₆ClNO
MDL Number: MFCD05861372
CAS Number: 1171992-10-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR