3-[(4-Methylbenzyl)oxy]piperidine hydrochloride

Supplier: Matrix Scientific

023165-500MG
101815-466EA 315.38 USD
101815-466
3-[(4-Methylbenzyl)oxy]piperidine hydrochloride
3-[(4-Methylbenzyl)oxy]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₃H₂₀ClNO
MDL Number: MFCD12405041
CAS Number: 1185301-39-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR