3-([1,1'-Biphenyl]-4-ylmethoxy)piperidine hydrochloride

Supplier: Matrix Scientific

023164-500MG
101815-464EA 315.38 USD
101815-464
3-([1,1'-Biphenyl]-4-ylmethoxy)piperidine hydrochloride
3-([1,1'-Biphenyl]-4-ylmethoxy)piperidine hydrochloride

Hazard: Irritant

Formula: C₁₈H₂₂ClNO
MDL Number: MFCD11100494
CAS Number: 1184989-56-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR